Thermo Scientific Chemicals

Imipramine hydrochloride, Thermo Scientific Chemicals

Catalog number: J63723.14
25 g, Each
Thermo Scientific Chemicals

Imipramine hydrochloride, Thermo Scientific Chemicals

Catalog number: J63723.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J63723.14
also known as J63723-14
Price (USD)
Price: 229.00
Online price: 194.65
Your price:
Quantity
-

Chemical Identifiers

CAS
113-52-0
IUPAC Name
hydrogen (3-{2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-yl}propyl)dimethylamine chloride
Molecular Formula
C19H25ClN2
InChI Key
XZZXIYZZBJDEEP-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CN(C)CCCN1C2=CC=CC=C2CCC2=CC=CC=C12
Elemental Analysis
Nitrogen: 8.5 - 9.1%
Thin Layer Chromatography
≥ 99%
Elemental Analysis
Carbon: 70.6 - 73.5%
Solution Test
Turbidity (50 mg/ml): Clear
Appearance (Color)
White to off White Powder

Description

Imipramine hydrochloride is used as a serotonin uptake inhibitor. It mainly used in the treatment of major depression and enuresis (inability to control urination). It has also been evaluated for use in panic disorder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Imipramine hydrochloride is used as a serotonin uptake inhibitor. It mainly used in the treatment of major depression and enuresis (inability to control urination). It has also been evaluated for use in panic disorder.

Solubility
Soluble in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text