Thermo Scientific™

Sodium phosphate, 0.2M buffer soln., pH 7.2

1L, Each
Thermo Scientific™

Sodium phosphate, 0.2M buffer soln., pH 7.2

1L, Each
Quantity
Unit Size
Catalog number: J63816.K2
also known as J63816-K2
Price (USD)
105.00 / Each
Your Price:
Quantity
-

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Sodium phosphate is widely used in molecular biology, biochemistry, and chromatography. Useful in conjunction with Sodium Phosphate, Monobasic in the preparation of biological buffers. Used in many applications including the purification of antibodies.

Solubility
Fully miscible.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures