Dihydroergotamine methanesulfonate
Dihydroergotamine methanesulfonate
Dihydroergotamine methanesulfonate
Dihydroergotamine methanesulfonate
Thermo Scientific Chemicals

Dihydroergotamine methanesulfonate

CAS: 6190-39-2 | C34H41N5O8S | 679.79 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
Catalog number J63840.MC
also known as J63840-MC
Price (TWD)
-
Request A Quote
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS6190-39-2
IUPAC Name(2R,4R,7R)-N-[(1S,2S,4R,7S)-7-benzyl-2-hydroxy-4-methyl-5,8-dioxo-3-oxa-6,9-diazatricyclo[7.3.0.0²,⁶]dodecan-4-yl]-6-methyl-6,11-diazatetracyclo[7.6.1.0²,⁷.0¹²,¹⁶]hexadeca-1(16),9,12,14-tetraene-4-carboxamide; methanesulfonic acid
Molecular FormulaC34H41N5O8S
InChI KeyADYPXRFPBQGGAH-UMYZUSPBSA-N
SMILESCS(O)(=O)=O.CN1C[C@@H](C[C@H]2[C@H]1CC1=CNC3=CC=CC2=C13)C(=O)N[C@]1(C)O[C@@]2(O)[C@@H]3CCCN3C(=O)[C@H](CC3=CC=CC=C3)N2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥96.0% (HPLC)
Appearance (Color)White to pale yellow to pale red
Optical Rotation-17.0 to -23.0°
Loss on Drying≤2.0%
Agonist at vascular serotonin receptors; partial agonist at alpha-adrenergic and dopamine D2 receptors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Agonist at vascular serotonin receptors
  • Partial agonist at alpha-adrenergic and dopamine D2 receptors
RUO – Research Use Only