Cyclocytidine hydrochloride, 98+%, Thermo Scientific Chemicals
Cyclocytidine hydrochloride, 98+%, Thermo Scientific Chemicals
Cyclocytidine hydrochloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclocytidine hydrochloride, 98+%, Thermo Scientific Chemicals

Anti-tumor agent
Have Questions?
產品號碼Quantity
J63845.03
亦稱為 J63845-03
1 g
產品號碼 J63845.03
亦稱為 J63845-03
價格 (TWD)
-
Quantity:
1 g
Request bulk or custom format
規格
CAS10212-25-6
ChEBICHEBI:74843
Chemical Name or MaterialCyclocytidine hydrochloride
ColorWhite
EINECS Number233-515-6
檢視更多
Anti-tumor agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only