Nalpha-(p-Toluenesulfonyl)-DL-lysine chloromethyl ketone hydrochloride, 98%
Nalpha-(p-Toluenesulfonyl)-DL-lysine chloromethyl ketone hydrochloride, 98%
Nalpha-(p-Toluenesulfonyl)-DL-lysine chloromethyl ketone hydrochloride, 98%
Thermo Scientific Chemicals

Nalpha-(p-Toluenesulfonyl)-DL-lysine chloromethyl ketone hydrochloride, 98%

Inhibitor of papain and trypsin | CAS: 4238-41-9 | C14H22Cl2N2O3S | 369.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
100 mg
Catalog number J63959.MF
also known as J63959-MF
Price (EUR)
99,90
Each
Quantity:
50 mg
Request bulk or custom format
Price (EUR)
99,90
Each
Specifications
CAS4238-41-9
Chemical Name or MaterialNalpha-(p-Toluenesulfonyl)-DL-lysine chloromethyl ketone hydrochloride
ColorWhite
EINECS Number224-199-0
Formula Weight369.31
View more
Inhibitor of papain and trypsin

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only