Informational:Informational:This product is animal origin and can only be shipped within the United States
Thermo Scientific Chemicals

Fluocinolone acetonide, Thermo Scientific Chemicals

Catalog number: J63996.MC
100 mg, Each
Thermo Scientific Chemicals

Fluocinolone acetonide, Thermo Scientific Chemicals

Catalog number: J63996.MC
100 mg, Each
Quantity
Catalog number: J63996.MC
also known as J63996-MC
Price (USD)

Chemical Identifiers

CAS
67-73-2
IUPAC Name
(1S,2S,4R,8S,9S,11S,12R,13S,19S)-12,19-difluoro-11-hydroxy-8-(2-hydroxyacetyl)-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
Molecular Formula
C24H30F2O6
InChI Key
FEBLZLNTKCEFIT-VSXGLTOVSA-N
SMILES
CC1(C)O[C@@H]2C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
Appearance (Color)
White to off-white
Assay (unspecified)
≥98.0%
Infrared spectrum
Conforms
Form
Powder

Description

Fluocinolone acetonide is primarily used in dermatology to reduce skin inflammation and relieve itching. It acts as an inhibitor of tumor cells and it can regulate lipid metabolism by modulating gene expression. It can also inhibit the expression of VEGF (vascular endothelial growth factor) in the retinal pigment epithelial cell line due to its high glucocorticoid receptor affinity. Furthermore, it can inhibit TNF-α angiogenesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluocinolone acetonide is primarily used in dermatology to reduce skin inflammation and relieve itching. It acts as an inhibitor of tumor cells and it can regulate lipid metabolism by modulating gene expression. It can also inhibit the expression of VEGF (vascular endothelial growth factor) in the retinal pigment epithelial cell line due to its high glucocorticoid receptor affinity. Furthermore, it can inhibit TNF-α angiogenesis.

Solubility
Soluble in water (partly), DMSO, alcohol, chloroform and methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text