2-Chloro-11-(1-piperazinyl)dibenzo[b,f]-1,4-oxazepine, 98%
2-Chloro-11-(1-piperazinyl)dibenzo[b,f]-1,4-oxazepine, 98%
2-Chloro-11-(1-piperazinyl)dibenzo[b,f]-1,4-oxazepine, 98%
Thermo Scientific Chemicals

2-Chloro-11-(1-piperazinyl)dibenzo[b,f]-1,4-oxazepine, 98%

A tricyclic norepinephrine uptake inhibitor | CAS: 14028-44-5 | C17H16ClN3O | 313.785 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number J64066.03
also known as J64066-03
Price (EUR)
206,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
206,00
Each
Specifications
CAS14028-44-5
ChEBICHEBI:2675
Chemical Name or Material2-Chloro-11-(1-piperazinyl)dibenzo[b,f]-1,4-oxazepine
ColorWhite
EINECS Number237-867-1
View more
A tricyclic norepinephrine uptake inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only