Thermo Scientific Chemicals

Cytidine-5'-diphosphocholine sodium salt

Catalog number: J64161.18
50 g, Each
Thermo Scientific Chemicals

Cytidine-5'-diphosphocholine sodium salt

Catalog number: J64161.18
50 g, Each
Quantity
Catalog number: J64161.18
also known as J64161-18
Price (USD)
Quantity
-

Chemical Identifiers

CAS
33818-15-4
IUPAC Name
sodium 2-(trimethylazaniumyl)ethyl ({[(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Molecular Formula
C14H25N4NaO11P2
InChI Key
YWAFNFGRBBBSPD-OCMLZEEQSA-M
SMILES
[Na+].C[N+](C)(C)CCOP([O-])(=O)OP([O-])(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=CC(N)=NC1=O
Assay (HPLC)
95.0% min.
Assay (unspecified)
Solubility: 5% w/v in water
Loss on Drying
6.0% max.

Description

Cytidine-5'-diphosphocholine sodium salt hydrate acts as a neuroprotective agent in situations of hypoxia and ischemia. It is used as biosynthetic intermediate of membrane phospholipids. It is a cerebral vasodilator and inhibits PLA2 activation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cytidine-5′-diphosphocholine sodium salt hydrate acts as a neuroprotective agent in situations of hypoxia and ischemia. It is used as biosynthetic intermediate of membrane phospholipids. It is a cerebral vasodilator and inhibits PLA2 activation.

Solubility
Soluble in water (100 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text