Informational:Informational:This product is animal origin and can only be shipped within the United States
Thermo Scientific Chemicals

Cortisone, Thermo Scientific Chemicals

Catalog number: J64182.06
5 g, Each
Thermo Scientific Chemicals

Cortisone, Thermo Scientific Chemicals

Catalog number: J64182.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J64182.06
also known as J64182-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
53-06-5
IUPAC Name
(1R,3aS,3bS,9aR,9bS,11aS)-1-hydroxy-1-(2-hydroxyacetyl)-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-7,10-dione
Molecular Formula
C21H28O5
InChI Key
MFYSYFVPBJMHGN-ZPOLXVRWSA-N
SMILES
C[C@]12CC(=O)[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO
Appearance (Color)
White
Form
Powder
Thin Layer Chromatography
98% min. (one spot)

Description

A steroid and immunosuppressant

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text