Thermo Scientific Chemicals

Benserazide hydrochloride, Thermo Scientific Chemicals

Catalog number: J64581.06
5 g, Each
Thermo Scientific Chemicals

Benserazide hydrochloride, Thermo Scientific Chemicals

Catalog number: J64581.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J64581.06
also known as J64581-06
Price (USD)
Price: 317.00
Online price: 273.65
Your price:
Quantity
-

Description

Inhibitor of L-aromatic amino acid decarboxylaseBenserazide hydrochloride is used as an antiparkinsonian. It is a peripheral decarboxylase inhibitor. It is used as an inhibitor for aromatic L-aminoacid decarboxylase. Further, it serves as an inhibitor of tyrosine hydroxylase (TH) and Tryptophan hydroxylase (TPH) inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Inhibitor of L-aromatic amino acid decarboxylaseBenserazide hydrochloride is used as an antiparkinsonian. It is a peripheral decarboxylase inhibitor. It is used as an inhibitor for aromatic L-aminoacid decarboxylase. Further, it serves as an inhibitor of tyrosine hydroxylase (TH) and Tryptophan hydroxylase (TPH) inhibitor.

Solubility
Soluble in water, dimethyl sulfoxide and methanol. Insoluble in ethanol and acetone.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text