Glycerol, ultrapure, 99.5+%
Glycerol, ultrapure, 99.5+%
Glycerol, ultrapure, 99.5+%
Thermo Scientific Chemicals

Glycerol, ultrapure, 99.5+%

Used in sample preparation and gel formation for polyacrylamide gel electrophoresis. Glycerol (5-10%) increases the density of a sample so that the sample will layer at the bottom of a gel’s sample well. | CAS: 56-81-5 | C3H8O3 | 92.09 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 mL
250 mL
500 mL
產品號碼 J64719.AK
亦稱為 J64719-AK
價格 (TWD)
-
Quantity:
250 mL
Request bulk or custom format
化學識別
CAS56-81-5
IUPAC Namepropane-1,2,3-triol
Molecular FormulaC3H8O3
InChI KeyPEDCQBHIVMGVHV-UHFFFAOYSA-N
SMILESOCC(O)CO
檢視更多
規格Specification Sheet規格表
Elemental AnalysisChloride: ≤ 10 ppm
Assay (unspecified)≥ 99.5%
Assay (unspecified)Sulfates: ≤ 20 ppm
Heavy Metals≤ 5 ppm as lead
Specific Gravity≥ 1.249

Glycerol, ultrapure is used in sample preparation and gel formation for polyacrylamide gel electrophoresis. Glycerol (5-10%) increases the density of a sample so that the sample will layer at the bottom of a gel's sample well. Glycerol is also used to aid in casting gradient gels and as a protein stabilizer and storage buffer component, glycerol is used as a solvent, plasticizer and humectant. Additional uses include as an aid in casting gradient gels, a protein stabilizer and storage buffer component.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only