uPA Inhibitor II, UK122
uPA Inhibitor II, UK122
Thermo Scientific Chemicals

uPA Inhibitor II, UK122

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
25 mg
Catalog number J64874.LB0
also known as J64874-#0
Price (EUR)
177,00
Each
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (EUR)
177,00
Each
Add to cart
Chemical Identifiers
CAS1186653-73-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormSolid
Assay from Suppliers CofA≥98.0% (Proton-NMR)

A cell-permeable oxazolidinone-carboxamidine compound that acts as a potent, selective, and active-site targeting inhibitor of urokinase-type plasminogen activatoruPA Inhibitor II, UK122 acts as a potent, selective and active-site targeting urokinase-type plasminogen activator (uPA) inhibitor. However, its activity against trypsin, thrombin, tPA, and plasmin is greatly reduced. It is also effectively inhibits the migration (80% inhibition) and invasion (68% inhibition) of uPA-expressing CFPAC-1 pancreatic cells, without affecting its proliferation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A cell-permeable oxazolidinone-carboxamidine compound that acts as a potent, selective, and active-site targeting inhibitor of urokinase-type plasminogen activator
RUO – Research Use Only
Contents & Storage
2°C to 8°C