4-Aminophthalhydrazide, 98%
4-Aminophthalhydrazide, 98%
4-Aminophthalhydrazide, 98%
Thermo Scientific Chemicals

4-Aminophthalhydrazide, 98%

A chemiluminescent molecule | CAS: 3682-14-2 | C8H7N3O2 | 177.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number J64921.03
also known as J64921-03
Price (EUR)
104,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
104,00
Each
Chemical Identifiers
CAS3682-14-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow
Infrared spectrumConforms
Purity≥97.5%
FormPowder
A chemiluminescent molecule

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only