2'-Fluoro-2'-deoxyinosine, 99%, Thermo Scientific Chemicals
2'-Fluoro-2'-deoxyinosine, 99%, Thermo Scientific Chemicals
2'-Fluoro-2'-deoxyinosine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2'-Fluoro-2'-deoxyinosine, 99%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
J65208.03
also known as J65208-03
1 g
Catalog number J65208.03
also known as J65208-03
Price (EUR)
394,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
394,00
Each
Specifications
CAS80049-87-2
Chemical Name or Material2'-Fluoro-2'-deoxyinosine
ColorWhite
Formula Weight270.22
IUPAC Name9-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only