Thermo Scientific Chemicals

JAK2 Inhibitor IV

Catalog number: J65506.LB0
5 mg, Each
Thermo Scientific Chemicals

JAK2 Inhibitor IV

Catalog number: J65506.LB0
5 mg, Each
Quantity
Catalog number: J65506.LB0
also known as J65506-#0
Price (USD)

Chemical Identifiers

CAS
1110502-30-1
Purity
≥95.0%
Form
Solid
Appearance (Color)
Pale yellow

Description

An aminoindazole compound that potently inhibits the activity of both the wild-type JAK2 and the active V617F mutant

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text