2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, 98%
2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, 98%
2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, 98%
Thermo Scientific Chemicals

2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, 98%

Chromogenic substrate for peroxidase assays | CAS: 30931-67-0 | C18H24N6O6S4 | 548.67 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
產品號碼 J65535.03
亦稱為 J65535-03
價格 (TWD)
-
Quantity:
1 g
Request bulk or custom format
化學識別
CAS30931-67-0
IUPAC Namediammonium (2Z)-3-ethyl-2-{2-[(2Z)-3-ethyl-6-sulfonato-2,3-dihydro-1,3-benzothiazol-2-ylidene]hydrazin-1-ylidene}-2,3-dihydro-1,3-benzothiazole-6-sulfonate
Molecular FormulaC18H24N6O6S4
InChI KeyOHDRQQURAXLVGJ-AXMZSLBLSA-N
SMILES[NH4+].[NH4+].CCN1C(SC2=CC(=CC=C12)S([O-])(=O)=O)=NN=C1/SC2=CC(=CC=C2N1CC)S([O-])(=O)=O
檢視更多
規格Specification Sheet規格表
Assay from Supplier's CofA≥97.5% (HPLC)
Water Content (Karl Fischer Titration)≤2.0%
Solution TestComplete: Clear pale green solution (0.5 gm in 1 ml of water)
UV/VisAbsorbance at 436nm: ≤ 0.25%
UV/VisAbsorbance at 748nm: ≤ 0.25%
檢視更多
Chromogenic substrate for peroxidase assays.2,2'-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid) diammonium salt is widely utilized as a peroxidase substrate suitable for use in enzyme-linked immunosorbent assay (ELISA) procedures. It plays an important role in the food industry to measure the antioxidant capacities of foods. It is used to observe the reaction kinetics of specific enzymes and involved in glucose estimating reagent in blood serum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chromogenic substrate for peroxidase assays.2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid) diammonium salt is widely utilized as a peroxidase substrate suitable for use in enzyme-linked immunosorbent assay (ELISA) procedures. It plays an important role in the food industry to measure the antioxidant capacities of foods. It is used to observe the reaction kinetics of specific enzymes and involved in glucose estimating reagent in blood serum.

Solubility
Dissolve in water at 50mg/ml. Solution may be slightly hazy.

Notes
Light Sensitive. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Garaguso, I.; Nardini, M. Polyphenols content, phenolics profile and antioxidant activity of organic red wines produced without sulfur dioxide/sulfites addition in comparison to conventional red wines. Food Chem. 2015, 179, 336-342.
  2. Wu, X.; Hu, Y.; Jin, J.; Zhou, N.; Wu, P.; Zhang, H.; Cai, C. Electrochemical Approach for Detection of Extracellular Oxygen Released from Erythrocytes Based on Graphene Film Integrated with Laccase and 2,2-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid). Anal. Chem. 2010, 82 (9), 3588-3596.