Thermo Scientific Chemicals

Deoxyribonucleic acid sodium salt, salmon testes, Thermo Scientific Chemicals

Catalog number: J65753.03
1 g, Each
Thermo Scientific Chemicals

Deoxyribonucleic acid sodium salt, salmon testes, Thermo Scientific Chemicals

Catalog number: J65753.03
1 g, Each
Quantity
Catalog number: J65753.03
also known as J65753-03
Price (USD)
Quantity
-

Chemical Identifiers

CAS
68938-01-2
IUPAC Name
Deoxyribonucleic acid sodium salt
SMILES
*
Molecular Weight (g/mol)
50000-100000
Synonym
DNA sodium salt
Appearance (Color)
Off-white
Form
Solid
Assay (unspecified)
E260,1%: 155.0 min.
Assay (unspecified)
N:P ratio: 1.5 to 1.8

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text