Alcohol dehydrogenase, yeast
Thermo Scientific Chemicals

Alcohol dehydrogenase, yeast

Have Questions?
Change viewbuttonViewtableView
Quantity:
75 kilounits
30 kilounit
150 kilounits
Catalog number J65869.VEX
also known as J65869-V!
Price (EUR)
178,00
Each
Quantity:
75 kilounits
Request bulk or custom format
Price (EUR)
178,00
Each
Chemical Identifiers
CAS9031-72-5
SpecificationsSpecification SheetSpecification Sheet
FormPowder
AssayProtein content (per mg): 150-350 microgram
AssaySpecific Activity: ≥290 U/mg protein
Assay (unspecified)Protein content (per mg): 150-350 microgram
Assay (unspecified)Specific Activity: ≥290 U/mg protein

Alcohol dehydrogenase catalyzes the reaction: RCH2OH +NAD+ ↔ RCHO + NADH + H+ It facilitates the interconversion between alcohols and aldehydes or ketones with the reduction of nicotinamide adenine dinucleotide (NAD+ to NADH). In biotransformation, alcohol dehydrogenases are often used for the synthesis of enantiomerically pure stereoisomers of chiral alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

How many grams of Alcohol dehydrogenase, yeast (Cat. No. J65869.9PL, J65869.VEX, J65869.WEX) are equivalent to the kilounit quantities listed on the website?

Here are the kilounit to gram conversions for the different catalog numbers of Alcohol dehydrogenase, yeast:
- Cat. No. J65869.9PL: 30 kilounits = 100 g
- Cat. No.J65869.VEX: 75 kilounits = 250 g
- Cat. No.J65869.WEX: 150 kilounits = 500 g