Alcohol dehydrogenase, yeast - FAQs
Alcohol dehydrogenase, yeast - FAQs
View additional product information for Alcohol dehydrogenase, yeast - FAQs (J65869.VEX, J65869.WEX, J65869.9PL)
1 product FAQs found
How many grams of Alcohol dehydrogenase, yeast (Cat. No. J65869.9PL, J65869.VEX, J65869.WEX) are equivalent to the kilounit quantities listed on the website?
Here are the kilounit to gram conversions for the different catalog numbers of Alcohol dehydrogenase, yeast:
- Cat. No. J65869.9PL: 30 kilounits = 100 g
- Cat. No.J65869.VEX: 75 kilounits = 250 g
- Cat. No.J65869.WEX: 150 kilounits = 500 g