Azido-PEG3-maleimide Kit - FAQs
Azido-PEG3-maleimide Kit - FAQs
View additional product information for Azido-PEG3-maleimide Kit - FAQs (J65984.MB, J65984.MC)
2 product FAQs found
What storage conditions should I use for the Azido-PEG3-maleimide Kit?
The Azido-PEG3-maleimide Kit should be stored in the dark at -20°C.
What is the protocol for Azide-PEG3-maleimide generation with the Azido-PEG3-maleimide Kit?
We recommend the following protocol for Azide-PEG3-maleimide generation with the Azido-PEG3-maleimide Kit:
  1. For the 25 mg kit, add 1 mL of a dry water-miscible solvent (like DMF or DMSO) to the azido-PEG-amine and shake for approximately 30 seconds to ensure mixing. For the 100 mg kit add 2.5 mL of solvent.
  2. Place the maleimide-NHS ester under nitrogen or dry atmosphere and slowly add the solution from step 1 with stirring or shaking.
  3. Stir or shake the reaction mixture for 30 minutes at room temperature. The progress of the reaction can be followed by Thin Layer Chromatograph.
    Note: For TLC, the solvent is typically methanol:methylene chloride (1:20), run on a silica gel normal phase plate and developed with a potassium permanganate spray. The Rf of the azide-PEG3-maleimide is slightly lower than the Rf for the maleimide-NHS ester. When the reaction is complete, it will be one clean spot on the plate.
  4. The reaction product will be ready-to-use azide-PEG3-maleimide.
The azide-PEG3-maleimide is present at ~ 75 mM if 1 mL of solvent is used in step 1. The 25 mg kit produces 0.075 mmol of azide-PEG3-maleimide.