Gatifloxacin sesquihydrate
Gatifloxacin sesquihydrate
Gatifloxacin sesquihydrate
Thermo Scientific Chemicals

Gatifloxacin sesquihydrate

A broad spectrum antibiotic | CAS: 180200-66-2 | C19H22FN3O4 | 375.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66036.06
also known as J66036-06
Price (EUR)
109,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
109,00
Each
Chemical Identifiers
CAS180200-66-2
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥98.0%
Water Content4.0-8.0%
FormPowder or crystals or crystalline powder
Residue on ignition≤0.5%
Appearance (Color)White to off-white
A broad spectrum antibiotic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only