Thermo Scientific Chemicals

Piperacillin, 95%, Thermo Scientific Chemicals

Catalog number: J66143.ME
500 mg, Each
Thermo Scientific Chemicals

Piperacillin, 95%, Thermo Scientific Chemicals

Catalog number: J66143.ME
500 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66143.ME
also known as J66143-ME
Price (USD)
Price: 138.00
Online price: 118.65
Your price:
Quantity
-

Chemical Identifiers

CAS
61477-96-1
IUPAC Name
sodium (2R,5S,6S)-6-[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Formula
C23H26N5NaO7S
InChI Key
WCMIIGXFCMNQDS-RAMASXNTSA-M
SMILES
[Na+].CCN1CCN(C(=O)N[C@@H](C(=O)N[C@@H]2[C@@H]3SC(C)(C)[C@H](N3C2=O)C([O-])=O)C2=CC=CC=C2)C(=O)C1=O
Appearance (Color)
White
Optical Rotation
+155.0 to +175.0°
Assay from Supplier's CofA
≥94.0%
Water Content
2.0-4.0%
Form
Powder or crystals or crystalline powder

Description

A semisynthetic penicillin with wide spectrum of antimicrobial activity, particularly pseudomonas strains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A semisynthetic penicillin with wide spectrum of antimicrobial activity, particularly pseudomonas strains.

Solubility
Freely soluble in methanol. Only sparingly soluble in aqueous solution at 0.119 mg/mL

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text