BQ-123
BQ-123
BQ-123
Thermo Scientific Chemicals

BQ-123

For cellular and molecular biology applications
Have Questions?
變更視圖buttonViewtableView
Quantity:
0.5 mg
1 mg
產品號碼 J66169.MCR
亦稱為 J66169-M^
價格 (TWD)
-
Quantity:
1 mg
Request bulk or custom format
規格
FormatSolid
Molecular Weight (g/mol)610.70
Product TypeBQ-123
Purification MethodHPLC
Quantity1 mg
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
內容物與存放
-30°C to -10°C