Thermo Scientific Chemicals

Glufosinate ammonium, 95%, Thermo Scientific Chemicals

Catalog number: J66186.MD
250 mg, Each
Thermo Scientific Chemicals

Glufosinate ammonium, 95%, Thermo Scientific Chemicals

Catalog number: J66186.MD
250 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66186.MD
also known as J66186-MD
Price (USD)
Price: 125.00
Online price: 106.65
Your price:
Quantity
-

Chemical Identifiers

CAS
77182-82-2
IUPAC Name
ammonium 2-amino-4-[hydroxy(methyl)phosphoryl]butanoate
Molecular Formula
C5H15N2O4P
InChI Key
ZBMRKNMTMPPMMK-UHFFFAOYNA-N
SMILES
[NH4+].CP(O)(=O)CCC(N)C([O-])=O
Form
Powder
Infrared spectrum
Conforms
Appearance (Color)
Cream to brown
Assay from Supplier's CofA
≥94.0%

Description

Used as broad spectrum post-emergence agrochemical for grapes, orchards, plantations, ornamentals, and non-cropland; Also used as crop desiccant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Glufosinate ammonium is a compound which exhibits broad-spectrum herbicidal properties through inhibition of glutamine synthetase, which disrupts cell membranes

Application

• Glufosinate ammonium acts as a non-selective herbicide

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text