Ethidium Bromide, Electrophoresis Reagent, 98% (dry weight)
Ethidium Bromide, Electrophoresis Reagent, 98% (dry weight)
Ethidium Bromide, Electrophoresis Reagent, 98% (dry weight)
Thermo Scientific Chemicals

Ethidium Bromide, Electrophoresis Reagent, 98% (dry weight)

CAS: 1239-45-8 | C21H20BrN3, C21H20BrN3 | 394.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number J66192.06
also known as J66192-06
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1239-45-8
IUPAC Name3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
Molecular FormulaC21H20BrN3
InChI KeyZMMJGEGLRURXTF-UHFFFAOYSA-N
SMILES[Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Titration)>97.5%
Infrared spectrumConforms
Loss on Drying5.0% max.
Solution(1% in water): clear, red
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only