Thermo Scientific Chemicals

Agarose, Genetic Technology Grade

Catalog number: J66208.14
25 g, Each
Thermo Scientific Chemicals

Agarose, Genetic Technology Grade

Catalog number: J66208.14
25 g, Each
Quantity
Catalog number: J66208.14
also known as J66208-14
Price (USD)
Price: 150.00
Online price: 130.65
Your price:
Quantity
-

Chemical Identifiers

CAS
9012-36-6
IUPAC Name
(2S,3R,4S,5R,6R)-2-{[(1S,3S,4S,5S,8R)-3-{[(2S,3R,4S,5S,6R)-2-{[(1S,3R,4S,5S,8R)-3,4-dihydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy}-4-hydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Formula
C24H38O19
InChI Key
MJQHZNBUODTQTK-WKGBVCLCSA-N
SMILES
OC[C@H]1O[C@@H](O[C@@H]2[C@@H]3CO[C@H]2[C@H](O)[C@H](O[C@H]2[C@@H](O)[C@@H](CO)O[C@@H](O[C@@H]4[C@@H]5CO[C@H]4[C@H](O)[C@H](O)O5)[C@@H]2O)O3)[C@H](O)[C@@H](O)[C@H]1O
Assay (unspecified)
Electroendosmosis: 0.09-0.13
Assay (unspecified)
Gel Strength (1%): 1200g/cm2 min.
Assay (unspecified)
Gelling temperature: 36?C ? 1.5
Water Content (Karl Fischer Titration)
10.0% max.

Description

Engineered to permit recovery of active DNA after separation, with minimal inhibition of restriction and ligation enzymes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Engineered to permit recovery of active DNA after separation, with minimal inhibition of restriction and ligation enzymes
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text