Galanin, Human
Thermo Scientific Chemicals

Galanin, Human

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 mg
1 mg
Catalog number J66251.MCR
also known as J66251-M^
Price (EUR)
268,00
Each
Quantity:
1 mg
Request bulk or custom format
Price (EUR)
268,00
Each
Specifications
FormatPowder
Molecular Weight (g/mol)3157.41
Product TypeGalanin, human
Purification MethodHPLC
Quantity1 mg
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C