D-Galacturonic acid monohydrate, 97%
D-Galacturonic acid monohydrate, 97%
D-Galacturonic acid monohydrate, 97%
Thermo Scientific Chemicals

D-Galacturonic acid monohydrate, 97%

CAS: 91510-62-2 | C6H9O7
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number J66282.14
also known as J66282-14
Price (EUR)
122,65
Online Exclusive
136,00
Save 13,35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
122,65
Online Exclusive
136,00
Save 13,35 (10%)
Each
Chemical Identifiers
CAS91510-62-2
SMILES*
Molecular Weight (g/mol)0.00
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or crystals or crystalline powder
Water Content5.0-12.0%
Purity≥96.0%
D-(+)-Galacturonic acid monohydrate is a chemical used in the synthesis of N-(D-galacturonoyl) amino acids and dipeptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-Galacturonic acid monohydrate is a chemical used in the synthesis of N-(D-galacturonoyl) amino acids and dipeptides.

Solubility
Soluble in water

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. Chang-Won Cho.; Do-Youn Lee.; Chan-Wha Kim. Concentration and purification of soluble pectin from mandarin peels using crossflow microfiltration system. Carbohydrate Polymers. 2003, 54, (1), 21-26.
  2. Verdini, R.A.; Zorrilla, S.E.; Rubiolo, A.C. Calcium Uptake during Immersion of Strawberries in CaCl2 Solutions. Journal of Food Science. 2008, 73, (7), C533-C539.