1,2,3,4-Tetra-O-acetyl-beta-D-glucopyranose, 98%
1,2,3,4-Tetra-O-acetyl-beta-D-glucopyranose, 98%
1,2,3,4-Tetra-O-acetyl-beta-D-glucopyranose, 98%
Thermo Scientific Chemicals

1,2,3,4-Tetra-O-acetyl-beta-D-glucopyranose, 98%

CAS: 13100-46-4 | C14H20O10 | 348.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
Catalog number J66307.ME
also known as J66307-ME
Price (USD)
86.65
Online Exclusive
96.60
Save 9.95 (10%)
Each
Quantity:
500 mg
Request bulk or custom format
Price (USD)
86.65
Online Exclusive
96.60
Save 9.95 (10%)
Each
Specifications
CAS13100-46-4
Chemical Name or Material1,2,3,4-Tetra-O-acetyl-beta-D-glucopyranose
ColorWhite
EINECS Number236-018-2
Formula Weight348.3
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only