Flumequine, 98%
Flumequine, 98%
Flumequine, 98%
Thermo Scientific Chemicals

Flumequine, 98%

Inhibits topoisomerases in gram negative bacteria | CAS: 42835-25-6 | C14H12FNO3 | 261.252 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66332.03
also known as J66332-03
Price (TWD)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Beilstein490724
CAS42835-25-6
ChEBICHEBI:85269
Chemical Name or MaterialFlumequine
EINECS Number255-962-6
View more
Inhibits topoisomerases in Gram-negative bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only