Cecropin P1, Porcine
Thermo Scientific Chemicals

Cecropin P1, Porcine

For cellular and molecular biology applications
Have Questions?
Catalog NumberQuantity
J66344.MCR
also known as J66344-M^
1 mg
Catalog number J66344.MCR
also known as J66344-M^
Price (TWD)
-
Quantity:
1 mg
Request bulk or custom format
Specifications
FormatLyophilized Powder
Molecular Weight (g/mol)3338.85
Product TypeCecropin P1, porcine
Purification MethodHPLC
Quantity1 mg
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C