Amyloid beta (1-42), Human
Thermo Scientific Chemicals

Amyloid beta (1-42), Human

For cellular and molecular biology applications | 4514.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 mg
1 mg
Catalog number J66387.EXD
also known as J66387-!D
Price (TWD)
-
Quantity:
0.5 mg
Request bulk or custom format
Chemical Identifiers
CAS107761-42-2
SpecificationsSpecification SheetSpecification Sheet
FormLyophilized powder
Molecular Weight4514.10 g/mol (theoretical): Consistent
Appearance (Color)White
Assay from Supplier's CofA≥94.0% (HPLC)
Aβ(s) peptides, their peptide fragments and mutated fragments are used to study a wide range of metabolic and regulatory functions including activation of kinases, regulation of cholesterol transport, function as a transcription factor, and regulators of inflammation. Aβ(s) peptides and their peptide fragments are also used to study oxidative stress, metal binding and mechanisms of protein cross-linking in the context of diseases such as Alzheimer’s disease and neurodegeneration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C