alpha-Mating Factor, yeast
Thermo Scientific Chemicals

alpha-Mating Factor, yeast

 
Have Questions?
Catalog NumberQuantity
J66502.MCR
also known as J66502-M^
1 mg
Catalog number J66502.MCR
also known as J66502-M^
Price (EUR)
93,90
Each
Quantity:
1 mg
Request bulk or custom format
Price (EUR)
93,90
Each
Specifications
CAS59401-28-4
FormatSolid
Quantity1 mg
Unit SizeEach
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
Store at -20°C