Protein Kinase C-epsilon Translocation Inhibitor Peptide, Negative Control
Protein Kinase C-epsilon Translocation Inhibitor Peptide, Negative Control
Thermo Scientific Chemicals

Protein Kinase C-epsilon Translocation Inhibitor Peptide, Negative Control

For cellular and molecular biology applications
Have Questions?
Catalog NumberQuantity
J66521.MCR
also known as J66521-M^
1 mg
Catalog number J66521.MCR
also known as J66521-M^
Price (TWD)
-
Request A Quote
Quantity:
1 mg
Request bulk or custom format
Specifications
For Use With (Application)A scrambled peptide with an identical amino acid composition to that of Protein Kinase C epsilon translocation Inhibitor Peptide. A cell impermeable negative control for PKC translocation inhibition.
FormatSolid
Molecular Weight (g/mol)843.98
Product TypeProtein Kinase C-epsilon Translocation Inhibitor Peptide, Negative Control
Purification MethodHPLC
View more
A scrambled peptide with an identical amino acid composition to that of Protein Kinase C epsilon translocation Inhibitor Peptide. A cell impermeable negative control for PKC translocation inhibition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C