Propidium iodide, 1mg/ml aqueous soln.
Propidium iodide, 1mg/ml aqueous soln.
Propidium iodide, 1mg/ml aqueous soln.
Thermo Scientific Chemicals

Propidium iodide, 1mg/ml aqueous soln.

CAS: 25535-16-4 | C27H34I2N4 | 668.406 g/mol
Have Questions?
Catalog NumberQuantity
J66584.AB
also known as J66584-AB
10 mL
Catalog number J66584.AB
also known as J66584-AB
Price (EUR)
107,65
Online Exclusive
119,00
Save 11,35 (10%)
Each
Quantity:
10 mL
Request bulk or custom format
Price (EUR)
107,65
Online Exclusive
119,00
Save 11,35 (10%)
Each
Chemical Identifiers
CAS25535-16-4
IUPAC Name3,8-diamino-5-{3-[diethyl(methyl)azaniumyl]propyl}-6-phenylphenanthridin-5-ium diiodide
Molecular FormulaC27H34I2N4
InChI KeyXJMOSONTPMZWPB-UHFFFAOYSA-M
SMILES[I-].[I-].CC[N+](C)(CC)CCC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Orange to red
lambda-max (UV-Vis)493 ± 3 nm
Assay (HPLC)≥ 95%
Thin Layer Chromatography≥ 95%
Membrane impermeant, phenanthridinium intercalator dye excited by mercury or xenon-arc lamps or argon-ion laser. Selectively stains dead cells and a counter stain for multicolor fluorescent images.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Membrane impermeant, phenanthridinium intercalator dye excited by mercury or xenon-arc lamps or argon-ion laser. Selectively stains dead cells and a counter stain for multicolor fluorescent images.

Solubility
Diffic
RUO – Research Use Only

General References:

  1. Jeffrey D. Macklis.; Roger D. Madison. Progressive incorporation of propidium iodide in cultured mouse neurons correlates with declining electrophysiological status: a fluorescence scale of membrane integrity. Journal of Neuroscience Methods. 1990, 31, (1), 43-46.
  2. Chang-Jing G. Yeh.; Bae-Li Hsi.; W. Page Faulk. Propidium iodide as a nuclear marker in immunofluorescence. II. Use with cellular identification and viability studies. Journal of Immunological Methods. 1981, 43, (3), 269-275.

Frequently asked questions (FAQs)

If I use propidium iodide (PI) for measuring cell viability before extracting DNA to perform qPCR and sequencing, will the PI interfere with the qPCR and sequencing reaction?

We recommend removing propidium iodide (PI) prior to PCR and sequencing. PI can be removed from the DNA by ethanol precipitation or spin column-based DNA purification that includes an ethanol wash step.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.