beta-D-Gentiobiose, 98%
beta-D-Gentiobiose, 98%
beta-D-Gentiobiose, 98%
Thermo Scientific Chemicals

beta-D-Gentiobiose, 98%

CAS: 554-91-6 | C12H22O11
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
1 g
Catalog number J66750.03
also known as J66750-03
Price (EUR)
263,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
263,00
Each
Chemical Identifiers
CAS554-91-6
IUPAC Name(2R,3R,4S,5S,6R)-6-({[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxane-2,3,4,5-tetrol
Molecular FormulaC12H22O11
InChI KeyDLRVVLDZNNYCBX-LIZSDCNHSA-N
SMILESOC[C@H]1O[C@@H](OC[C@H]2O[C@@H](O)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or crystalline powder
Assay (HPLC)≥98.0%
Water Content (Karl Fischer Titration)≤0.5%
Melting point180 193°C
View more
Gentiobiose was used in binding assays to determine if coelomic cytolytic factor 1 (CCF-1) binds to β-1,3-glucans to stimulate the pro-PO-activating system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Gentiobiose was used in binding assays to determine if coelomic cytolytic factor 1 (CCF-1) binds to β-1,3-glucans to stimulate the pro-PO-activating system.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only
Markus Weishaupt.; Steffen Eller.; Peter H. Seeberger. Chapter Twenty-Two - Solid Phase Synthesis of Oligosaccharides. Methods in Enzymology. 2010, 478, 463-484.