PACAP (1-27), Human, Ovine, Rat
Thermo Scientific Chemicals

PACAP (1-27), Human, Ovine, Rat

For cellular and molecular biology applications
Have Questions?
Catalog NumberQuantity
J66752.MCR
also known as J66752-M^
1 mg
Catalog number J66752.MCR
also known as J66752-M^
Price (TWD)
-
Quantity:
1 mg
Request bulk or custom format
Specifications
FormatSolid
Molecular Weight (g/mol)3147.64
Product TypePACAP (1-27), human, ovine, rat
Purification MethodHPLC
Quantity1 mg
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C