Head Activator Neuropeptide
Thermo Scientific Chemicals

Head Activator Neuropeptide

 
Have Questions?
Catalog NumberQuantity
J66870.MCR
also known as J66870-M^
1 mg
Catalog number J66870.MCR
also known as J66870-M^
Price (EUR)
54,90
Each
Quantity:
1 mg
Request bulk or custom format
Price (EUR)
54,90
Each
Specifications
Common NameHydra Peptide
FormatSolid
Molecular Weight (g/mol)1125.50
Product TypeHead Activator Neuropeptide
Purification MethodHPLC
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C