Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys
Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys
Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys
Thermo Scientific Chemicals

Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys

For cellular and molecular biology applications
Have Questions?
Catalog NumberQuantity
J66909.MCR
also known as J66909-M^
1 mg
Catalog number J66909.MCR
also known as J66909-M^
Price (EUR)
75,60
Each
Quantity:
1 mg
Request bulk or custom format
Price (EUR)
75,60
Each
Specifications
Common NameFlag PeptideÖ Sigma-Aldrich
FormatLyophilized Powder
Molecular Weight (g/mol)1013.00
Product TypeAsp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys
Purification MethodHPLC
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C