Thermo Scientific Chemicals

1,2,3,4,6-Penta-O-acetyl-D-mannopyranose, 98%

Catalog number: J66973.22
100 g, Each
Thermo Scientific Chemicals

1,2,3,4,6-Penta-O-acetyl-D-mannopyranose, 98%

Catalog number: J66973.22
100 g, Each
Quantity
Catalog number: J66973.22
also known as J66973-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25941-03-1
IUPAC Name
[3,4,5,6-tetrakis(acetyloxy)oxan-2-yl]methyl acetate
Molecular Formula
C16H22O11
InChI Key
LPTITAGPBXDDGR-UHFFFAOYNA-N
SMILES
CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
Assay (unspecified)
Specific rotation: +51 to +57?C, c=1 in chloroform
Assay (unspecified)
>97.5%

Description

1,2,3,4,6-Penta-O-acetyl-D-mannopyranose is used for the preparation of glycopyranoside phosphates for the study of T-lymphocyte mediated inflammatory diseases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4,6-Penta-O-acetyl-D-mannopyranose is used for the preparation of glycopyranoside phosphates for the study of T-lymphocyte mediated inflammatory diseases.

Solubility
Soluble in dichloromethane, ether, ethyl acetate, methanol, and chloroform.

Notes
Keep container tightly closed in a dry and well-ventilated place. Store in cool place. Store at -20°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text