L-Alanyl-L-glutamine, 99%
L-Alanyl-L-glutamine, 99%
L-Alanyl-L-glutamine, 99%
Thermo Scientific Chemicals

L-Alanyl-L-glutamine, 99%

L-Alanyl-L-glutamine, CAS # 39537-23-0, is a synthesized dipeptide, which can be digested to form stable L-glutamine in vitro. | CAS: 39537-23-0 | C8H15N3O4 | 217.23 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 J66996.22
亦稱為 J66996-22
價格 (TWD)
-
Quantity:
100 g
Request bulk or custom format
化學識別
CAS39537-23-0
IUPAC Name(2S)-2-[(2S)-2-aminopropanamido]-4-carbamoylbutanoic acid
Molecular FormulaC8H15N3O4
InChI KeyHJCMDXDYPOUFDY-WHFBIAKZSA-N
SMILESC[C@H](N)C(=O)N[C@@H](CCC(N)=O)C(O)=O
檢視更多
規格Specification Sheet規格表
Assay (HPLC)>98.5%
Loss on Drying0.5% maximum
A substitute for glutamine in mammalian cell culture media; stable to heat-sterilization.L-Alanyl-L-glutamine is used as a component in pharmaceuticals, cosmetics and food products. It is a stable Gln dipeptide, which protects mice against the jejunal crypt depletion in the setting of dietary protein and fat deficiency. It provides nutritional requirements, improves intestinal health and enhances the immune system. It is also involved in various clinical studies such as cancer treatment, radiation and chemotherapy. It finds application in biotechnology as a component for mammalian cell culture medium. It improves pancreatic beta-cell function, maintaining the mitochondrial metabolism and stimulus-secretion coupling essential for insulin release.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• L-Alanyl-L-glutamine is a synthetic dipeptide that degrades to L-glutamine

Application

• L-Alanyl-L-glutamine can be used as a substitute for glutamine in mammalian cell culture media and is stable to heat-sterilization
• It is a stable Gln dipeptide, which protects mice against the jejunal crypt depletion in the setting of dietary protein and fat deficiency
• It improves pancreatic beta-cell function, maintaining the mitochondrial metabolism and stimulus-secretion coupling essential for insulin release
• When this dipeptide is added to cultures of antibody-producing CHO cells in vitro, it reduces apoptosis and promotes antibody production

RUO – Research Use Only

General References:

  1. Nova, M. V.; Gonçalves, M. D. C. P.; Nogueira, A. C.; Herculano, L. D. S.; Medina, A. N.; Bazotte, R. B.; Bruschi, M. L. Formulation and characterization of ethylcellulose microparticles containing. l-alanyl-l-glutamine peptide. Drug Dev. Ind. Pharm. 2014, 40(10), 1308-1317.
  2. Wang, G.; Wang, Y.; Zhang, J.; Luan, Q.; Ma, Y.; Hao, H. Modeling and simulation of thermodynamic properties of L-Alanyl-L-Glutamine in different solvents. Ind. Eng. Chem. Res. 2014, 53 (8), 3385-3392.