Specifications
CAS1405-10-3
Chemical Name or MaterialNeomycin sulfate
EINECS Number215-773-1
Health Hazard 1H317-H334-H361d
Health Hazard 2GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
View more