Thermo Scientific Chemicals

Fumagillin, 95%, Thermo Scientific Chemicals

Catalog number: J67045.MCR
1 mg, Each
Thermo Scientific Chemicals

Fumagillin, 95%, Thermo Scientific Chemicals

Catalog number: J67045.MCR
1 mg, Each
Quantity
Catalog number: J67045.MCR
also known as J67045-M^
Price (USD)

Chemical Identifiers

CAS
23110-15-8
IUPAC Name
(2E,4E,6E,8E)-10-{[(3R,4S,5S,6R)-5-methoxy-4-[(2R)-2-methyl-3-(3-methylbut-2-en-1-yl)oxiran-2-yl]-1-oxaspiro[2.5]octan-6-yl]oxy}-10-oxodeca-2,4,6,8-tetraenoic acid
Molecular Formula
C26H34O7
InChI Key
NGGMYCMLYOUNGM-IJIYPQOHSA-N
SMILES
CO[C@@H]1[C@@H](CC[C@]2(CO2)[C@H]1[C@@]1(C)OC1CC=C(C)C)OC(=O)C=CC=CC=CC=CC(O)=O
Form
Solid
Assay from Supplier's CofA
≥94.0%
Appearance (Color)
White to pale yellow or pale cream

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text