6-Methoxy-2-naphthylacetic acid, 99%
6-Methoxy-2-naphthylacetic acid, 99%
6-Methoxy-2-naphthylacetic acid, 99%
6-Methoxy-2-naphthylacetic acid, 99%
Thermo Scientific Chemicals

6-Methoxy-2-naphthylacetic acid, 99%

CAS: 23981-47-7 | C13H12O3 | 216.24 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 mg
10 mg
50 mg
產品號碼 J67138.MA
亦稱為 J67138-MA
價格 (TWD)
-
申請報價
Quantity:
10 mg
Request bulk or custom format
規格
CAS23981-47-7
ChEBICHEBI:35628
Chemical Name or Material6-Methoxy-2-naphthylacetic acid
EINECS Number245-967-1
Formula Weight216.24
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only