Silver nanoparticles applications include molecular diagnostics and photonic devices, which take advantage of the novel optical properties of these nanomaterials. An increasingly common application is the use of silver nanoparticles for antimicrobial coatings, and many textiles, keyboards, wound dressings, and biomedical devices now contain silver nanoparticles that continuously release a low level of silver ions to provide protection against bacteria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Silver nanoparticles applications include molecular diagnostics and photonic devices, which take advantage of the novel optical properties of these nanomaterials. An increasingly common application is the use of silver nanoparticles for antimicrobial coatings, and many textiles, keyboards, wound dressings, and biomedical devices now contain silver nanoparticles that continuously release a low level of silver ions to provide protection against bacteria.
Solubility
Fully miscible in water.
Notes
Peak SPR: 425-450nm. 1.7E+10 nanoparticles/ml.
RUO – Research Use Only
JJ Mock.; M Barbic.; DR Smith.; DA Schultz. Shape effects in plasmon resonance of individual colloidal silver nanoparticles.J. Chem. Phys.,2002,116,(6), 6755-6777.
S Link.; ZL Wang.; MA El-Sayed. Alloy formation of gold-silver nanoparticles and the dependence of the plasmon absorption on their composition.J. Phys. Chem. 1999,103,(18), 3529-3533.