Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane hydrochloride, Molecular Biology Grade, 99+%

Catalog number: J67233.22
100 g, Each
Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane hydrochloride, Molecular Biology Grade, 99+%

Catalog number: J67233.22
100 g, Each
Quantity
Catalog number: J67233.22
also known as J67233-22
Price (USD)
Price: 58.00
Online price: 49.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1185-53-1
IUPAC Name
hydrogen 2-amino-2-(hydroxymethyl)propane-1,3-diol chloride
Molecular Formula
C4H12ClNO3
InChI Key
QKNYBSVHEMOAJP-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NC(CO)(CO)CO
Impurity content
Calcium, Ca: ≤5ppm
Protease activity
None detected
Impurity content
Manganese, Mn: ≤1ppm
Impurity content
Zinc, Zn: ≤1ppm
Residual water
≤0.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • For adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols
  • Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text