Gold nanoparticles, 80nm, supplied in 0.1mM PBS, 95%, OD1, 553 nm absorption
Gold nanoparticles, 80nm, supplied in 0.1mM PBS, 95%, OD1, 553 nm absorption
Gold nanoparticles, 80nm, supplied in 0.1mM PBS, 95%, OD1, 553 nm absorption
Thermo Scientific Chemicals

Gold nanoparticles, 80nm, supplied in 0.1mM PBS, 95%, OD1, 553 nm absorption

CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 mL
100 mL
產品號碼 J67285.AC
亦稱為 J67285-AC
價格 (TWD)
-
Quantity:
25 mL
Request bulk or custom format
化學識別
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
檢視更多
規格Specification Sheet規格表
Comment~7.82E+9
CommentBuffer: 0.1 mM Phosphate-Buffered Saline (0.1X PBS)
Core size80nm (±3nm)
For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM
RUO – Research Use Only