Gold nanoparticles, 60nm, supplied in 0.1mg/mL sodium citrate with stabilizer, OD1, 540nm absorption, Thermo Scientific Chemicals
Gold nanoparticles, 60nm, supplied in 0.1mg/mL sodium citrate with stabilizer, OD1, 540nm absorption, Thermo Scientific Chemicals
Gold nanoparticles, 60nm, supplied in 0.1mg/mL sodium citrate with stabilizer, OD1, 540nm absorption, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gold nanoparticles, 60nm, supplied in 0.1mg/mL sodium citrate with stabilizer, OD1, 540nm absorption, Thermo Scientific Chemicals

CAS: 7440-57-5 | Molecular Formula: Au | Molecular Weight: 196.97 g/mol
Have Questions?
Catalog NumberQuantity
J67306.AC
also known as J67306-AC
25 mL
Catalog number J67306.AC
also known as J67306-AC
Price (USD)
-
Quantity:
25 mL
Request bulk or custom format
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Buffer0.1 mg/mL citrate supplemented with a proprietary stabilizing solution
CommentCore size: 60nm ± 3nm
CommentMean hydrodynamic diameter (nm): 71.0
CommentParticles/mL: ~1.96E±10
View more

For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols
  • Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications
RUO – Research Use Only

Frequently asked questions (FAQs)

My Gold nanoparticles that are supplied in 0.1 mg/ml sodium citrate with stabilizer have turned a clear to bluish color. What causes this?

Irreversible aggregation is characterized by a clear to bluish solution upon the addition of salt. Since non-functionalized gold nanoparticles are sensitive to salt-containing buffers, re-suspension should always be performed in ultra-pure water to prevent irreversible aggregation.

How can I wash Gold nanoparticles, supplied in 0.1 mg/ml sodium citrate with stabilizer?

Although it is not generally necessary to wash the gold nanoparticles prior to use, some applications may require additional washing procedures. The easiest way to remove possible contaminants in the nanoparticles solution is by centrifugation. Centrifugation force is dependent on size of the gold nanoparticles. We recommend the following to wash gold nanoparticles:
1. Place 1 mL aliquot of colloidal gold in a 1.5 mL microcentrifuge tube.
2. Centrifuge the gold nanoparticles for 30 minutes using the recommended G force based on particle size (see below).
3. Remove the supernatant and re-suspend in an appropriate volume of ultra-pure water.
4. Vortex to re-disperse the particles.

Centrifugation speed guide for nanoparticle size:
size - G force
5 nm - 100,000 g
10 nm - 17,000 g
15 nm - 17,000 g
20 nm - 6,500 g
30 nm - 4,500 g
40 nm - 2,500 g
50 nm - 2,000 g
60 nm - 1,125 g
80 nm - 600 g
100 nm - 400 g

Do I need to wash Gold nanoparticles that are supplied in 0.1 mg/ml sodium citrate with stabilizer before use?

Although it is not generally necessary to wash the gold nanoparticles prior to use, some applications may require additional washing procedures. The easiest way to remove possible contaminants in the nanoparticles solution is by centrifugation. Centrifugation force is dependent on size of the gold nanoparticles. We recommend the following to wash gold nanoparticles:
1. Place 1 mL aliquot of colloidal gold in a 1.5 mL microcentrifuge tube.
2. Centrifuge the gold nanoparticles for 30 minutes using the recommended G force based on particle size (see below).
3. Remove the supernatant and re-suspend in an appropriate volume of ultra-pure water.
4. Vortex to re-disperse the particles.

Centrifugation speed guide for nanoparticle size:
size - G force
5 nm - 100,000 g
10 nm - 17,000 g
15 nm - 17,000 g
20 nm - 6,500 g
30 nm - 4,500 g
40 nm - 2,500 g
50 nm - 2,000 g
60 nm - 1,125 g
80 nm - 600 g
100 nm - 400 g

What are the recommended storage conditions for the Gold nanoparticles that are supplied in 0.1 mg/ml sodium citrate with stabilizer?

We recommend 4°C storage for our Gold nanoparticles that are supplied in 0.1 mg/ml sodium citrate with stabilizer. Do not freeze the particles because this causes irreversible aggregation. When stored for a long period of time, gold nanoparticles may sediment at the bottom of the flask, which is especially true for larger particle sizes. Prior to use, re-suspend the sedimented particles by swirling until a homogenous solution is obtained.