Thermo Scientific Chemicals

beta-Lapachone, 98%

Catalog number: J67308.LB0
5 mg, Each
Thermo Scientific Chemicals

beta-Lapachone, 98%

Catalog number: J67308.LB0
5 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J67308.LB0
also known as J67308-0
Price (USD)
Price: 159.00
Online price: 136.65
Your price:
Quantity
-

Chemical Identifiers

CAS
4707-32-8
IUPAC Name
2,2-dimethyl-2H,3H,4H,5H,6H-naphtho[1,2-b]pyran-5,6-dione
Molecular Formula
C15H14O3
InChI Key
QZPQTZZNNJUOLS-UHFFFAOYSA-N
SMILES
CC1(C)CCC2=C(O1)C1=CC=CC=C1C(=O)C2=O
Form
Solid
Purity
≥97.5%
Appearance (Color)
Orange to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text