Manoalide, 98%, Thermo Scientific Chemicals
Manoalide, 98%, Thermo Scientific Chemicals
Manoalide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Manoalide, 98%, Thermo Scientific Chemicals

 
Have Questions?
產品號碼Quantity
J67341.MCR
亦稱為 J67341-M^
1 mg
產品號碼 J67341.MCR
亦稱為 J67341-M^
價格 (TWD)
-
Quantity:
1 mg
Request bulk or custom format
規格
CAS75088-80-1
ChEBICHEBI:66666
Chemical Name or MaterialManoalide
ColorColorless to White
Formula Weight416.56
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only